missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat PCSK1 / NEC1 Protein
A DNA sequence encoding the Rattus norvegicus (Rat) PCSK1 / NEC1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9322-ye-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_058787.1 | |
PCSK1 / NEC1 Protein | |
Research Use Only | |
Pcsk1 | |
Recombinant Protein | |
Yeast | |
SVPRDSALNL FNDPMWNQQW YLQDTRMTAS LPKLDLHVIP VWQKGITGKG VVITVLDDGL EWNHTDIYAN YDPEASYDFN DNDHDPFPRY DPTNENKHGT RCAGEIAMQA NNHKCGVGVA YNSKVGGIRM LDGIVTDAIE ASSIGFNPGH VDIYSASWGP NDDGKTVEGP GRLAQKAFEY GVKQGRQGKG SIFVWASGNG GRQGDNCDCD GYTDSIYTIS ISSASQQGLS PWYAEKCSST LATSYSSGDY TDQRITSADL HNDCTETHTG TSASAPLAAG IFALALEANP NLTWRDMQHL VVWTSEYDPL ANNPGWKKNG AGLMVNSRFG FGLLNAKALV DLADPRTWRN VPEKKECIIK DNNFEPRALK ANGEVIVEIP TRACEGQENA INSLEHVQFE ATIEYSRRGD LHVTLTSAAG TSTVLLAERE RDTSPNGFKN WDFMSVHTWG ENPVGTWTLK VTDMSGRMQN EGRIVNWKLI LHGTSSQPEH MKQPRVYTSY NTVQNDRRGV EKMVNVVEEK PTQNSLNGNL LVPKNSSSSS VEDRRDEQVQ GAPSKAMLRL LQSAFSKNTP SKQSSKIPSA KLSVPYEGLY EALEKLNKPS QLEDSEDSLY SDYVDVFYNT KPYKHRDDRL LQALMDILNE KN | |
Greater than 90% as determined by SDS-PAGE. |
25204 | |
200 μg | |
Not determined. | |
Full Length | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |