missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Nitric oxide synthase, endothelial Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Nitric oxide synthase, endothelial, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8865-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_068610.1 | |
Nitric oxide synthase, endothelial Protein | |
Research Use Only | |
Nos3 | |
Recombinant Protein | |
E. coli | |
ATILYGSETG RAQSYAQQLG RLFRKAFDPR VLCMDEYDVV SLEHEALVLV VTSTFGNGDP PENGESFAAA LMEMSGPYNS SPRPEQHKSY KIRFNSVSCS DPLVSSWRRK RKESSNTDSA GALGTLRFCV FGLGSRAYPH FCAFARAVDT RLEELGGERL LQLGQGDELC GQ | |
Greater than 90% as determined by SDS-PAGE. |
24600 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |