missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat MMP-2 / CLG4A Protein

A DNA sequence encoding the Rattus norvegicus (Rat) MMP-2 / CLG4A Protein, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6369-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16022810

  • 1925.00 EUR / 500µg
Estimated Shipment: 02-02-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_112316.2
MMP-2 / CLG4A Protein
500 μg
Not determined.
Full Length
Rat
His
Tris-based buffer, 50% glycerol
81686
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Mmp2
Recombinant Protein
Yeast
YNFFPRKPKW DKNQITYRII GYTPDLDPET VDDAFARALK VWSDVTPLRF SRIHDGEADI MINFGRWEHG DGYPFDGKDG LLAHAFAPGT GVGGDSHFDD DELWTLGEGQ VVRVKYGNAD GEYCKFPFLF NGREYSSCTD TGRSDGFLWC STTYNFEKDG KYGFCPHEAL FTMGGNGDGQ PCKFPFRFQG TSYNSCTTEG RTDGYRWCGT TEDYDRDKKY GFCPETAMST VGGNSEGAPC VFPFTFLGNK YESCTSAGRS DGKVWCATTT NYDDDRKWGF CPDQGYSLFL VAAHEFGHAM GLEHSQDPGA LMAPIYTYTK NFRLSNDDIK GIQELYGPSP DADTDTGTGP TPTLGPVTPE ICKQDIVFDG IAQIRGEIFF FKDRFIWRTV TPRDKPTGPL LVATFWPELP EKIDAVYEAP QEEKAVFFAG NEYWVYSAST LERGYPKPLT SLGLPPDVQQ VDAAFNWSKN KKTYIFSGDK FWRYNEVKKK MDPGFPKLIA DSWNAIPDNL DAVVDLQGGG HSYFFKGAYY LKLENQSLKS VKFGSIKSDW LGC
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers