missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat Kynurenine 3-monooxygenase Protein

A DNA sequence encoding the Rattus norvegicus (Rat) Kynurenine 3-monooxygenase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6272-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16082100

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_067604.1
Kynurenine 3-monooxygenase Protein
Research Use Only
KMO
Recombinant Protein
E. coli
MASSDTEGKR VVVIGGGLVG ALNACFLAKR NFQVDVYEAR EDIRVANFMR GRSINLALSY RGRQALKAVG LEDQIVSKGV PMKARMIHSL SGKKSAIPYG NKSQYILSIS REKLNKDLLT AVESYPNAKV HFGHKLSKCC PEEGILTMLG PNKVPRDITC DLIVGCDGAY STVRAHLMKK PRFDYSQQYI PHGYMELTIP PKNGEYAMEP NCLHIWPRNA FMMIALPNMD KSFTCTLFMS FEEFEKLPTH SDVLDFFQKN FPDAIPLMGE QALMRDFFLL PAQPMISVKC SPFHLKSRCV LMGDAAHAIV PFFGQGMNAG FEDCLVFDEL MDKFNNDLSV CLPEFSRFRI PDDHAISDLS MYNYIEMRAH VNSRWFLFQR LLDK
Greater than 90% as determined by SDS-PAGE.
59113
50 μg
Not determined.
Partial (see sequence information for more details).
Rat
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers