missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Kynurenine 3-monooxygenase Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Kynurenine 3-monooxygenase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6272-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_067604.1 | |
Kynurenine 3-monooxygenase Protein | |
Research Use Only | |
KMO | |
Recombinant Protein | |
E. coli | |
MASSDTEGKR VVVIGGGLVG ALNACFLAKR NFQVDVYEAR EDIRVANFMR GRSINLALSY RGRQALKAVG LEDQIVSKGV PMKARMIHSL SGKKSAIPYG NKSQYILSIS REKLNKDLLT AVESYPNAKV HFGHKLSKCC PEEGILTMLG PNKVPRDITC DLIVGCDGAY STVRAHLMKK PRFDYSQQYI PHGYMELTIP PKNGEYAMEP NCLHIWPRNA FMMIALPNMD KSFTCTLFMS FEEFEKLPTH SDVLDFFQKN FPDAIPLMGE QALMRDFFLL PAQPMISVKC SPFHLKSRCV LMGDAAHAIV PFFGQGMNAG FEDCLVFDEL MDKFNNDLSV CLPEFSRFRI PDDHAISDLS MYNYIEMRAH VNSRWFLFQR LLDK | |
Greater than 90% as determined by SDS-PAGE. |
59113 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Rat | |
His-SUMO | |
Tris-based buffer, 50% glycerol |