missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat KNG1 / BDK / kininogen-1 Protein

A DNA sequence encoding the Rattus norvegicus (Rat) KNG1 / BDK / kininogen-1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9238-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16007901

  • 1705.00 EUR / 500µg


Estimated Shipment
24-02-2020
Add to basket

Specifications

Specifications

NP_036873.1
KNG1 / BDK / kininogen-1 Protein
500 μg
Not determined.
Partial (see sequence information for more details).
Rat
His
Tris-based buffer, 50% glycerol
25087
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Kng1
Recombinant Protein
Yeast
APRVKKPKES TTVSPSYIAR VQEERDPGNE QGPIHGHGWL HAKQIKNKNH QGHKHGHGIG HGHQKPHGLG HGHQLKLDDL KQQREDGYDH RHPVGHGHGQ RHGHGHGHGH GRDKHTNKDK NNVKHTDQRR APLTSSSEDN TTSTQIQGRT EGFTLNPPLA QPAVISRGFQ DSGFTEGVIA TTSPYDTETH DDLIPDIHVQ PDSLSFKLIS DFPEATSHKC PGRPWKPVSR KDPTIETTEF SDFDLLDALS
Videos
Documents