missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat IL-18 / IL-1F4 Protein

A DNA sequence encoding the Rattus norvegicus (Rat) IL-18 / IL-1F4, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8901-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16017091

  • 2985.00 EUR / 1mg
Estimated Shipment: 28-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_062038.1
IL-18 / IL-1F4 Protein
Research Use Only
IL18
Recombinant Protein
E. coli
HFGRLHCTTA VIRSINDQVL FVDKRNPPVF EDMPDIDRTA NESQTRLIIY MYKDSEVRGL AVTLSVKDGR MSTLSCKNKI ISFEEMNPPE NIDDIKSDLI FFQKRVPGHN KMEFESSLYE GHFLACQKED DAFKLVLKRK DENGDKSVMF TLTNLHQS
Greater than 90% as determined by SDS-PAGE.
29197
1 mg
Not determined.
Full Length
Rat
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers