missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat IFNG / Interferon Gamma Protein

A DNA sequence encoding the Rattus norvegicus (Rat) IFNG / Interferon Gamma Protein, was expressed in the hosts and tags indicated.

245.00 EUR - 2645.00 EUR

Specifications

Accession Number NP_620235.1
Gene ID (Entrez) 25712
Name IFNG / Interferon Gamma Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16066761
View Documents Promotion Details
enQuireBio™
QP8840-EC-10UG
10 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16006771
View Documents Promotion Details
enQuireBio™
QP8840-EC-50UG
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16056761
View Documents Promotion Details
enQuireBio™
QP8840-EC-100UG
100 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16086761
View Documents Promotion Details
enQuireBio™
QP8840-EC-200UG
200 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16096761
View Documents Promotion Details
enQuireBio™
QP8840-EC-500UG
500 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16076761
View Documents Promotion Details
enQuireBio™
QP8840-EC-1MG
1 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Specifications

NP_620235.1
IFNG / Interferon Gamma Protein
Not determined.
Full Length
Rat
GST
Tris-based buffer, 50% glycerol
25712
Research Use Only
IFNG
Recombinant Protein
E. coli
QGTLIESLES LKNYFNSSSM DAMEGKSLLL DIWRNWQKDG NTKILESQII SFYLRLFEVL KDNQAISNNI SVIESHLITN FFSNSKAKKD AFMSIAKFEV NNPQIQHKAV NELIRVIHQL SPESSLRKRK RSRC
Greater than 90% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications