missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat HRAS / GTPase Hras Protein

A DNA sequence encoding the Rattus norvegicus (Rat) HRAS / GTPase Hras, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8837-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16006751

  • 985.00 EUR / 200µg


Estimated Shipment
23-03-2020
Add to basket

Specifications

Specifications

NP_001091711.1
HRAS / GTPase Hras Protein
200 μg
Not determined.
Full Length
Rat
His
Tris-based buffer, 50% glycerol
293621
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Hras
Recombinant Protein
E. coli
TEYKLVVVGA GGVGKSALTI QLIQNHFVDE YDPTIEDSYR KQVVIDGETC LLDILDTAGQ EEYSAMRDQY MRTGEGFLCV FAINNTKSFE DIHQYREQIK RVKDSDDVPM VLVGNKCDLA ARTVESRQAQ DLARSYGIPY IETSAKTRQG VEDAFYTLVR EIRQHKLRKL NPPDESGPGC MSCKC
Videos
Documents