missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat BDNF Protein Isoform 2 Protein
A DNA sequence encoding the Rattus norvegicus (Rat) BDNF Protein Isoform 2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8869-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001257559.1 | |
BDNF Protein Isoform 2 Protein | |
Research Use Only | |
BDNF | |
Recombinant Protein | |
E. coli | |
RRGELSVCDS ISEWVTAADK KTAVDMSGGT VTVLEKVPVS KGQLKQYFYE TKCNPMGYTK EGCRGIDKRH WNSQCRTTQS YVRALTMDSK KRIGWRFIRI DTSCVCTL | |
Greater than 90% as determined by SDS-PAGE. |
24225 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |