missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Angiopoietin 1 / ANG1 / ANGPT1 Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Angiopoietin 1 / ANG1 / ANGPT1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8975-ye-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_445998.1 | |
Angiopoietin 1 / ANG1 / ANGPT1 Protein | |
Research Use Only | |
Angpt1 | |
Recombinant Protein | |
Yeast | |
SNQRRSPENG GRRYNRIQHG QCAYTFILPE HDGNCRESAT EQYNTNALQR DAPHVETDFS SQKLQHLEHV MENYTQWLQK LENYIVENMK SEMAQIQQNA VQNHTATMLE IGTSLLSQTA EQTRKLTDVE TQVLNQTSRL EIQLLENSLS TYELEKQLLQ QTNEILKIQE KNSLLEHKIL EMEGKHKEEL DTLKEEKENL QGLVTRQTFI IQELEKQLSR ATSNNSVLQK QQLELMDTVH NLVSLCTKEV LLKGGKREEE KPFRDCADVY QAGFNKSGIY TIYFNNMPEP KKVFCNMDVN EGGWTVIQHR EDGSLDFQRG WKEYKMGFGN PSGEYWLGNE FIFAITSQRQ YMLRIELMDW EGNRAYSQYD RFHIGNQKQN YRLYLKGHTG TAGKQSSLIL HGADFSTKDA DNDNCMCKCA LMLTGGWWFD ACGPSNLNGM FYTAGQNHGK LNGIKWHYFK GPSYSLRSTT MMIRPLDF | |
Greater than 90% as determined by SDS-PAGE. |
89807 | |
100 μg | |
Not determined. | |
Full Length | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |