missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant RABV PhosphoProtein
A DNA sequence encoding the Rabies virus (strain PM) (RABV) Phosphoprotein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6952-1mg
Additional Details : Weight : 0.01000kg
Specifications
RABV PhosphoProtein | |
Research Use Only | |
P | |
Recombinant Protein | |
E. coli | |
MSKIFVNPSA IRAGLADLEM AEETVDLINR NIEDNQAHLQ GEPIEVDNLP EDMGRLHLDD GKSPNPGEMA KVGEGKYRED FQMDEGEDPS FLFQSYLENV GVQIVRQMRS GERFLKIWSQ TVEEIISYVA VNFPNPPGKS SEDKSTQTTG RELKKETTPT PSQRESQSSK ARMAAQTASG PPALEWSATN EKDDLSVEAE IAHQIAESFS KKYKFPSRSS GILLYNFEQL KMNLDDIVKE AKNVPGVTRL AHDGSKLPLR CVLGWVALAN SKKFQLLVES DKLSKIMQDD LNRYTSC | |
Greater than 90% as determined by SDS-PAGE. |
1 mg | |
Not determined. | |
Full Length | |
RABV | |
His | |
Tris-based buffer, 50% glycerol |