missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant RABV GlycoProtein

A DNA sequence encoding the Rabies virus (strain PM) (RABV) Glycoprotein, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7413-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16048750

  • 2985.00 EUR / 1mg
Estimated Shipment: 31-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

RABV GlycoProtein
Research Use Only
G
Recombinant Protein
E. coli
KFPIYTILDK LGPWSPIDIH HLSCPNNLVV EDEGCTNLSG FSYMELKVGY ILAIKMNGFT CTGVVTEAET YTNFVGYVTT TFKRKHFRPT PDACRAAYNW KMAGDPRYEE SLHNPYPDYR WLRTVKTTKE SLVIISPSVA DLDPYDRSLH SRVFPSGKCS GVAVSSTYCS TNHDYTIWMP ENPRLGMSCD IFTNSRGKRA SKGSETCGFV DERGLYKSLK GACKLKLCGV LGLRLMDGTW VAMQTSNETK WCPPDQLVNL HDFRSDEIEH LVVEELVRKR EECLDALESI MTTKSVSFRR LSHLRKLVPG FGKAYTIFNK TLMEADAHYK SVRTWNEILP SKGCLRVGGR CHPHVNGVFF NGIILGPDGN VLIPEMQSSL LQQHMELLES SVIPLVHPLA DPSTVFKDGD EAEDFVEVHL PDVHNQVSGV DLGLPNWGKY
Greater than 90% as determined by SDS-PAGE.
1 mg
Not determined.
Full Length
RABV
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers