missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rabbit Tissue factor pathway inhibitor Protein

A DNA sequence encoding the Oryctolagus cuniculus (Rabbit) Tissue factor pathway inhibitor, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6775-ye-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16005760

Quantity Price
1 352.00 EUR / 10µg
6 328.49 EUR / 10µg
Estimated Shipment
31-08-2020
Add to basket


Specifications

Specifications

NP_001095184.1
Tissue factor pathway inhibitor Protein
10 μg
Not determined.
Full Length
Rabbit
His
Tris-based buffer, 50% glycerol
100009401
Greater than 90% as determined by SDS-PAGE.
Research Use Only
TFPI
Recombinant Protein
Yeast
AAEEDEEFTN ITDIKPPLQK PTHSFCAMKV DDGPCRAYIK RFFFNILTHQ CEEFIYGGCE GNENRFESLE ECKEKCARDY PKMTTKLTFQ KGKPDFCFLE EDPGICRGYI TRYFYNNQSK QCERFKYGGC LGNLNNFESL EECKNTCENP TSDFQVDDHR TQLNTVNNTL INQPTKAPRR WAFHGPSWCL PPADRGLCQA NEIRFFYNAI IGKCRPFKYS GCGGNENNFT SKKACITACK KGFIPKSIKG GLIKTKRKKK KQPVKITYVE TFVKKT
Videos
Documents