missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rabbit Tissue factor pathway inhibitor Protein
A DNA sequence encoding the Oryctolagus cuniculus (Rabbit) Tissue factor pathway inhibitor, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6775-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001095184.1 | |
Tissue factor pathway inhibitor Protein | |
Research Use Only | |
TFPI | |
Recombinant Protein | |
E. coli | |
AAEEDEEFTN ITDIKPPLQK PTHSFCAMKV DDGPCRAYIK RFFFNILTHQ CEEFIYGGCE GNENRFESLE ECKEKCARDY PKMTTKLTFQ KGKPDFCFLE EDPGICRGYI TRYFYNNQSK QCERFKYGGC LGNLNNFESL EECKNTCENP TSDFQVDDHR TQLNTVNNTL INQPTKAPRR WAFHGPSWCL PPADRGLCQA NEIRFFYNAI IGKCRPFKYS GCGGNENNFT SKKACITACK KGFIPKSIKG GLIKTKRKKK KQPVKITYVE TFVKKT | |
Greater than 90% as determined by SDS-PAGE. |
100009401 | |
500 μg | |
Not determined. | |
Full Length | |
Rabbit | |
His | |
Tris-based buffer, 50% glycerol |