missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rabbit Tissue factor pathway inhibitor Protein

A DNA sequence encoding the Oryctolagus cuniculus (Rabbit) Tissue factor pathway inhibitor, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6775-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16065750

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001095184.1
Tissue factor pathway inhibitor Protein
Research Use Only
TFPI
Recombinant Protein
E. coli
AAEEDEEFTN ITDIKPPLQK PTHSFCAMKV DDGPCRAYIK RFFFNILTHQ CEEFIYGGCE GNENRFESLE ECKEKCARDY PKMTTKLTFQ KGKPDFCFLE EDPGICRGYI TRYFYNNQSK QCERFKYGGC LGNLNNFESL EECKNTCENP TSDFQVDDHR TQLNTVNNTL INQPTKAPRR WAFHGPSWCL PPADRGLCQA NEIRFFYNAI IGKCRPFKYS GCGGNENNFT SKKACITACK KGFIPKSIKG GLIKTKRKKK KQPVKITYVE TFVKKT
Greater than 90% as determined by SDS-PAGE.
100009401
200 μg
Not determined.
Full Length
Rabbit
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers