missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Pig Uricase Protein

A DNA sequence encoding the Sus scrofa (Pig) Uricase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6871-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16036440

  • 860.00 EUR / 100µg
Estimated Shipment: 30-01-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_999435.1
Uricase Protein
100 μg
Not determined.
Full Length
Pig
His-SUMO
Tris-based buffer, 50% glycerol
397510
Greater than 90% as determined by SDS-PAGE.
Research Use Only
UOX
Recombinant Protein
E. coli
MAHYRNDYKK NDEVEFVRTG YGKDMIKVLH IQRDGKYHSI KEVATSVQLT LSSKKDYLHG DNSDVIPTDT IKNTVNVLAK FKGIKSIETF AVTICEHFLS SFKHVIRAQV YVEEVPWKRF EKNGVKHVHA FIYTPTGTHF CEVEQIRNGP PVIHSGIKDL KVLKTTQSGF EGFIKDQFTT LPEVKDRCFA TQVYCKWRYH QGRDVDFEAT WDTVRSIVLQ KFAGPYDKGE YSPSVQKTLY DIQVLTLGQV PEIEDMEISL PNIHYLNIDM SKMGLINKEE VLLPLDNPYG RITGTVKRKL TSRL
Videos
SDS
Documents
Certificates
Special Offers

Special Offers