missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Pig E-Selectin / CD62e / SELE Protein
A DNA sequence encoding the Sus scrofa (Pig) E-Selectin / CD62e / SELE, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6661-ye-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_999433.1 | |
E-Selectin / CD62e / SELE Protein | |
Research Use Only | |
SELE | |
Recombinant Protein | |
Yeast | |
WSYSASTETM TFDDASAYCQ QRYTHLVAIQ NHAEIEYLNS TFNYSASYYW IGIRKINGTW TWIGTKKALT PEATNWAPGE PNNKQSNEDC VEIYIKRDKD SGKWNDERCS KKKLALCYTA ACTPTSCSGH GECIETINSS TCQCYPGFRG LQCEQVVECD ALENPVNGVV TCPQSLPWNT TCAFECKEGF ELIGPEHLQC TSSGSWDGKK PTCKAVTCDT VGHPQNGDVS CNHSSIGEFA YKSTCHFTCA EGFGLQGPAQ IECTAQGQWT QQAPVCKAVK CPAVSQPKNG LVKFTHSPTG EFTYKSSCAF SCEEGFELRG SAQLACTSQG QWTQEVPSCQ VVQCSSLEVP REINMSCSGE PVFGAVCTFA CPEGWMLNGS VALTCGATGH WSGMLPTCEA PAESKIP | |
Greater than 90% as determined by SDS-PAGE. |
397508 | |
200 μg | |
Not determined. | |
Extracellular Domain | |
Pig | |
His | |
Tris-based buffer, 50% glycerol |