missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant P. gingivalis Gingipain R2 Protein

A DNA sequence encoding the Porphyromonas gingivalis (strain ATCC BAA-308 / W83) Gingipain R2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6989-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16046930

  • 940.00 EUR / 100µg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

WP_010956050.1
100 μg
Not determined.
Partial (see sequence information for more details).
P. gingivalis
His-SUMO
Tris-based buffer, 50% glycerol
P. gingivalis Gingipain R2 Protein
Research Use Only
rgpB
Recombinant Protein
E. coli
YTPVEEKENG RMIVIVPKKY EEDIEDFVDW KNQRGLRTEV KVAEDIASPV TANAIQQFVK QEYEKEGNDL TYVLLVGDHK DIPAKITPGI KSDQVYGQIV GNDHYNEVFI GRFSCESKED LKTQIDRTIH YERNITTEDK WLGQALCIAS AEGGPSADNG ESDIQHENII ANLLTQYGYT KIIKCYDPGV TPKNIIDAFN GGISLANYTG HGSETAWGTS HFGTTHVKQL TNSNQLPFIF DVAC
Greater than 90% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers