missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant P. gingivalis Gingipain R1 Protein
A DNA sequence encoding the Porphyromonas gingivalis Gingipain R1, was expressed in the hosts and tags indicated.
530.00 EUR - 1485.00 EUR
Specifications
Name | P. gingivalis Gingipain R1 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | rgpA |
Protein Length | Partial (see sequence information for more details). |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16037640
|
enQuireBio™
QP7192-MA-20UG |
20 μg |
530.00 EUR
20µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16027640
|
enQuireBio™
QP7192-MA-100UG |
100 μg |
1485.00 EUR
100µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16047640
|
enQuireBio™
QP7192-MA-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
P. gingivalis Gingipain R1 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
P. gingivalis | |
His | |
Tris-based buffer, 50% glycerol |
Research Use Only | |
rgpA | |
Recombinant Protein | |
Mammal | |
YTPVEEKQNG RMIVIVAKKY EGDIKDFVDW KNQRGLRTEV KVAEDIASPV TANAIQQFVK QEYEKEGNDL TYVLLVGDHK DIPAKITPGI KSDQVYGQIV GNDHYNEVFI GRFSCESKED LKTQIDRTIH YERNITTEDK WLGQALCIAS AEGGPSADNG ESDIQHENVI ANLLTQYGYT KIIKCYDPGV TPKNIIDAFN GGISLVNYTG HGSETAWGTS HFGTTHVKQL TNSNQLPFIF DVACVNGDFL FSMPCFAEAL MRAQKDGKPT GTVAIIASTI NQSWASPMRG QDEMNEILCE KHPNNIKRTF GGVTMNGMFA MVEKYKKDGE KMLDTWTVFG DPSLLVRTLV PTKMQVTAPA QINLTDASVN VSCDYNGAIA TISANGKMFG SAVVENGTAT INLTGLTNES TLTLTVVGYN KETVIKTINT NGEPNPYQPV SNLTATTQGQ KVTLKWDAPS TKTNATTNTA RSVDGIRELV LLSVSDAPEL LRS | |
Greater than 80% as determined by SDS-PAGE. |