missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant P. aeruginosa Regulatory protein RhlR Protein

A DNA sequence encoding the Pseudomonas aeruginosa Regulatory protein RhlR, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7228-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16027800

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_252167.1
P. aeruginosa Regulatory protein RhlR Protein
Research Use Only
rhlR
Recombinant Protein
E. coli
MRNDGGFLLW WDGLRSEMQP IHDSQGVFAV LEKEVRRLGF DYYAYGVRHT IPFTRPKTEV HGTYPKAWLE RYQMQNYGAV DPAILNGLRS SEMVVWSDSL FDQSRMLWNE ARDWGLCVGA TLPIRAPNNL LSVLSVARDQ QNISSFEREE IRLRLRCMIE LLTQKLTDLE HPMLMSNPVC LSHREREILQ WTADGKSSGE IAIILSISES TVNFHHKNIQ KKFDAPNKTL AAAYAAALGL I
Greater than 84.5% as determined by SDS-PAGE.
878968
10 μg
Not determined.
Full Length
P. aeruginosa
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers