missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant P. aeruginosa Beta-lactamase Protein

A DNA sequence encoding the Pseudomonas aeruginosa (strain ATCC 15692 / PAO1 / 1C / PRS 101 / LMG 12228) Beta-lactamase, was expressed in the hosts and tags indicated. option, ab

Brand:  enQuireBio™ QP7119-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16007430

  • 1375.00 EUR / 200µg


Estimated Shipment
07-08-2020
Add to basket

Specifications

Specifications

NP_252799.1
P. aeruginosa Beta-lactamase Protein
200 μg
Not determined.
Full Length
P. aeruginosa
His-SUMO
Tris-based buffer, 50% glycerol
878149
Greater than 90% as determined by SDS-PAGE.
Research Use Only
ampC
Recombinant Protein
E. coli
GEAPADRLKA LVDAAVQPVM KANDIPGLAV AISLKGEPHY FSYGLASKED GRRVTPETLF EIGSVSKTFT ATLAGYALTQ DKMRLDDRAS QHWPALQGSR FDGISLLDLA TYTAGGLPLQ FPDSVQKDQA QIRDYYRQWQ PTYAPGSQRL YSNPSIGLFG YLAARSLGQP FERLMEQQVF PALGLEQTHL DVPEAALAQY AQGYGKDDRP LRVGPGPLDA EGYGVKTSAA DLLRFVDANL HPERLDRPWA QALDATHRGY YKVGDMTQGL GWEAYDWPIS LKRLQAGNST PMALQPHRIA RLPAPQALEG QRLLNKTGST NGFGAYVAFV PGRDLGLVIL ANRNYPNAER VKIAYAILSG LEQQGKVPLK R
Videos
Documents