missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant P. aeruginosa Beta-lactamase OXA-10 Protein

A DNA sequence encoding the Pseudomonas aeruginosa Beta-lactamase OXA-10, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7059-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16047270

  • 555.00 EUR / 50µg
Estimated Shipment: 28-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

WP_000846390.1
50 μg
Not determined.
Full Length
P. aeruginosa
His-SUMO
Tris-based buffer, 50% glycerol
P. aeruginosa Beta-lactamase OXA-10 Protein
Research Use Only
bla
Recombinant Protein
E. coli
SITENTSWNK EFSAEAVNGV FVLCKSSSKS CATNDLARAS KEYLPASTFK IPNAIIGLET GVIKNEHQVF KWDGKPRAMK QWERDLTLRG AIQVSAVPVF QQIAREVGEV RMQKYLKKFS YGNQNISGGI DKFWLEGQLR ISAVNQVEFL ESLYLNKLSA SKENQLIVKE ALVTEAAPEY LVHSKTGFSG VGTESNPGVA WWVGWVEKET EVYFFAFNMD IDNESKLPLR KSIPTKIMES EGIIGG
Greater than 90% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers