missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mycoplasma Uncharacterized lipoprotein MPN_083 Protein
A DNA sequence encoding the Mycoplasma pneumoniae (strain ATCC 29342 / M129) Uncharacterized lipoprotein MPN_083, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6955-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_109771.1 | |
Mycoplasma Uncharacterized lipoprotein MPN_083 Protein | |
Research Use Only | |
MPN_083 | |
Recombinant Protein | |
E. coli | |
CSFKDYIPTP SFRKDFSTEN NFVKNKVPGK DDIYSKFYDL TFSLNFVNNQ AQEFGTGWLI DWKGDENKNL SKNKEGQTAS QTRSSSEQTT DQDANLFTAY IATNLHVADG LKNDQDYAPY NKDGWGQPYP YQQKTQSFLL GKYTKPNVQL VKTNYEKPED AVIEQKLKED SLLFIQTSTL PKTAYAAIDP VNFSYNPTRT NGFWTAGKYN VYNGGNSIGN Y | |
Greater than 90% as determined by SDS-PAGE. |
877109 | |
100 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Mycoplasma | |
His-SUMO | |
Tris-based buffer, 50% glycerol |