missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse VEGF-C Protein

A DNA sequence encoding the Mus musculus (Mouse) VEGF-C, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6880-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16036510

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_033532.1
VEGF-C Protein
Research Use Only
Vegfc
Recombinant Protein
E. coli
AHYNTEILKS IDNEWRKTQC MPREVCIDVG KEFGAATNTF FKPPCVSVYR CGGCCNSEGL QCMNTSTGYL SKTLFEITVP LSQGPKPVTI SFANHTSCRC MSKLDVYRQV HSIIRR
Greater than 90% as determined by SDS-PAGE.
22341
500 μg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers