missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Ubiquitin-protein ligase E3A Protein
A DNA sequence encoding the Mus musculus (Mouse) Ubiquitin-protein ligase E3A, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6865-ma-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_035798.2 | |
Ubiquitin-protein ligase E3A Protein | |
Research Use Only | |
Ube3a | |
Recombinant Protein | |
Mammal | |
HIIDDALVRL EMIAMENPAD LKKQLYVEFE GEQGVDEGGV SKEFFQLVVE EIFNPDIGMF TYDEATKLFW FNPSSFETEG QFTLIGILGL AIYNNCILDV HFPMVVYRKL MGKKGTFRDL GDSHPVLYQS LKDLLEYEGS VEDDMMITFQ ISQTDLFGNP MMYDLKENGD KIPITNENRK EFVISYSDYI LNKSVEKQFK AFRRGFHMVT NESPLKYLFR PEEIELLICG SRNLDFQALE ETTEYDGGYT RESVVIREFW EIVHSFTDEQ KRLFLLFTTG TDRAPVGGLG KLKMIIAKNG PDTERLPTSH TCFNVLLLPE YSSKEKLNV | |
Greater than 90% as determined by SDS-PAGE. |
22215 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
Flag | |
Tris-based buffer, 50% glycerol |