missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Tryptophan Hydroxylase 1 / TPH1 Protein
A DNA sequence encoding the Mus musculus (Mouse) Tryptophan Hydroxylase 1 / TPH1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6826-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001129556.1 | |
Tryptophan Hydroxylase 1 / TPH1 Protein | |
Research Use Only | |
Tph1 | |
Recombinant Protein | |
E. coli | |
MIEDNKENKE NKDHSSERGR VTLIFSLENE VGGLIKVLKI FQENHVSLLH IESRKSKQRN SEFEIFVDCD ISREQLNDIF PLLKSHATVL SVDSPDQLTA KEDVMETVPW FPKKISDLDF CANRVLLYGS ELDADHPGFK DNVYRRRRKY FAELAMNYKH GDPIPKIEFT EEEIKTWGTI FRELNKLYPT HACREYLRNL PLLSKYCGYR EDNIPQLEDV SNFLKERTGF SIRPVAGYLS PRDFLSGLAF RVFHCTQYVR HSSDPLYTPE PDTCHELLGH VPLLAEPSFA QFSQEIGLAS LGASEETVQK LATCYFFTVE FGLCKQDGQL RVFGAGLLSS ISELKHALSG HAKVKPFDPK IACKQECLIT SFQDVYFVSE SFEDAKEKMR EFAKTVKRPF GLKYNPYTQS VQVLRDTKSI TSAMNELRYD LDVISDALAR VTRWPSV | |
Greater than 90% as determined by SDS-PAGE. |
21990 | |
200 μg | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |