missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Tryptase Protein
A DNA sequence encoding the Mus musculus (Mouse) Tryptase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6831-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_112464.4 | |
Tryptase Protein | |
Research Use Only | |
Tpsab1 | |
Recombinant Protein | |
E. coli | |
AHGNKWPWQV SLRANDTYWM HFCGGSLIHP QWVLTAAHCV GPDVADPNKV RVQLRKQYLY YHDHLMTVSQ IITHPDFYIV QDGADIALLK LTNPVNISDY VHPVPLPPAS ETFPSGTLCW VTGWGNIDNG VNLPPPFPLK EVQVPIIENH LCDLKYHKGL ITGDNVHIVR DDMLCAGNEG HDSCQGDSGG PLVCKVEDTW LQAGVVSWGE GCAQPNRPGI YTRVTYYLDW IHH | |
Greater than 90% as determined by SDS-PAGE. |
100503895 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |