missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Tryptase Protein

A DNA sequence encoding the Mus musculus (Mouse) Tryptase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6831-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16016140

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_112464.4
Tryptase Protein
Research Use Only
Tpsab1
Recombinant Protein
E. coli
AHGNKWPWQV SLRANDTYWM HFCGGSLIHP QWVLTAAHCV GPDVADPNKV RVQLRKQYLY YHDHLMTVSQ IITHPDFYIV QDGADIALLK LTNPVNISDY VHPVPLPPAS ETFPSGTLCW VTGWGNIDNG VNLPPPFPLK EVQVPIIENH LCDLKYHKGL ITGDNVHIVR DDMLCAGNEG HDSCQGDSGG PLVCKVEDTW LQAGVVSWGE GCAQPNRPGI YTRVTYYLDW IHH
Greater than 90% as determined by SDS-PAGE.
100503895
500 μg
Not determined.
Partial (see sequence information for more details).
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers