missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse TPT1 / Tumor Protein

A DNA sequence encoding the Mus musculus (Mouse) TPT1 / Tumor, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6835-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16026170

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_033455.1
TPT1 / Tumor Protein
Research Use Only
Tpt1
Recombinant Protein
E. coli
MIIYRDLISH DELFSDIYKI REIADGLCLE VEGKMVSRTE GAIDDSLIGG NASAEGPEGE GTESTVVTGV DIVMNHHLQE TSFTKEAYKK YIKDYMKSLK GKLEEQKPER VKPFMTGAAE QIKHILANFN NYQFFIGENM NPDGMVALLD YREDGVTPFM IFFKDGLEME KC
Greater than 90% as determined by SDS-PAGE.
22070
50 μg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers