missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse TPT1 / Tumor Protein
A DNA sequence encoding the Mus musculus (Mouse) TPT1 / Tumor, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6835-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_033455.1 | |
TPT1 / Tumor Protein | |
Research Use Only | |
Tpt1 | |
Recombinant Protein | |
E. coli | |
MIIYRDLISH DELFSDIYKI REIADGLCLE VEGKMVSRTE GAIDDSLIGG NASAEGPEGE GTESTVVTGV DIVMNHHLQE TSFTKEAYKK YIKDYMKSLK GKLEEQKPER VKPFMTGAAE QIKHILANFN NYQFFIGENM NPDGMVALLD YREDGVTPFM IFFKDGLEME KC | |
Greater than 90% as determined by SDS-PAGE. |
22070 | |
500 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |