missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Toll-like receptor 7 Protein
A DNA sequence encoding the Mus musculus (Mouse) Toll-like receptor 7, was expressed in the hosts and tags indicated.
245.00 EUR - 2985.00 EUR
Specifications
Accession Number | NP_001277684.1 |
---|---|
Gene ID (Entrez) | 170743 |
Name | Toll-like receptor 7 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16015950
|
enQuireBio™
QP6801-EC-100UG |
100 μg |
675.00 EUR
100µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16035950
|
enQuireBio™
QP6801-EC-1MG |
1 mg |
2985.00 EUR
1mg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16025950
|
enQuireBio™
QP6801-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16045950
|
enQuireBio™
QP6801-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16055950
|
enQuireBio™
QP6801-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16065950
|
enQuireBio™
QP6801-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001277684.1 | |
Toll-like receptor 7 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
GST | |
Tris-based buffer, 50% glycerol |
170743 | |
Research Use Only | |
Tlr7 | |
Recombinant Protein | |
E. coli | |
FRWFPKTLPC EVKVNIPEAH VIVDCTDKHL TEIPEGIPTN TTNLTLTINH IPSISPDSFR RLNHLEEIDL RCNCVPVLLG SKANVCTKRL QIRPGSFSGL SDLKALYLDG NQLLEIPQDL PSSLHLLSLE ANNIFSITKE NLTELVNIET LYLGQNCYYR NPCNVSYSIE KDAFLVMRNL KVLSLKDNNV TAVPTTLPPN LLELYLYNNI IKKIQENDFN NLNELQVLDL SGNCPRCYNV PYPCTPCENN SPLQIHDNAF NSLTELKVLR LHSNSLQHVP PTWFKNMRNL QELDLSQNYL AREIEEAKFL HFLPNLVELD FS | |
Greater than 90% as determined by SDS-PAGE. |