missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse TNFRSF4 / OX40 / CD134 Protein

A DNA sequence encoding the Mus musculus (Mouse) TNFRSF4 / OX40 / CD134, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6816-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16006050

  • 985.00 EUR / 200µg


Estimated Shipment
20-02-2020
Add to basket

Specifications

Specifications

NP_035789.1
TNFRSF4 / OX40 / CD134 Protein
200 μg
Not determined.
Extracellular Domain
Mouse
His-SUMO
Tris-based buffer, 50% glycerol
22163
Greater than 90% as determined by SDS-PAGE.
Research Use Only
TNFRSF4
Recombinant Protein
E. coli
VTARRLNCVK HTYPSGHKCC RECQPGHGMV SRCDHTRDTL CHPCETGFYN EAVNYDTCKQ CTQCNHRSGS ELKQNCTPTQ DTVCRCRPGT QPRQDSGYKL GVDCVPCPPG HFSPGNNQAC KPWTNCTLSG KQTRHPASDS LDAVCEDRSL LATLLWETQR PTFRPTTVQS TTVWPRTSEL PSPPTLVTPE GP
Videos
Documents