missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse TIMP-1 / TIMP1 Protein

A DNA sequence encoding the Mus musculus (Mouse) TIMP-1 / TIMP1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6795-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16005910

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001037849.1
TIMP-1 / TIMP1 Protein
Research Use Only
Timp1
Recombinant Protein
E. coli
CSCAPPHPQT AFCNSDLVIR AKFMGSPEIN ETTLYQRYKI KMTKMLKGFK AVGNAADIRY AYTPVMESLC GYAHKSQNRS EEFLITGRLR NGNLHISACS FLVPWRTLSP AQQRAFSKTY SAGCGVCTVF PCLSIPCKLE SDTHCLWTDQ VLVGSEDYQS RHFACLPRNP GLCTWRSLGA R
Greater than 90% as determined by SDS-PAGE.
21857
10 μg
Not determined.
Full Length
Mouse
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers