missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse TECTB Protein

A DNA sequence encoding the Mus musculus (Mouse) TECTB, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9985-iv-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16039141

  • 1495.00 EUR / 100µg
Estimated Shipment: 02-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_033374.2
TECTB Protein
Research Use Only
Tectb
Recombinant Protein
E. coli
KSCTPNKADV ILVFCYPKTI ITKIPECPYG WEVHQLALGG LCYNGVHEGG YYQFVIPDLS PKNKSYCGTQ SEYKPPIYHF YSHIVSNDST VIVKNQPVNY SFSCTYHSTY LVNQAAFDQR VATVHVKNGS MGTFESQLSL NFYTNAKFST KKEAPFVLET SEIGSDLFAG VEAKGLSVRF KVVLNSCWAT PSADFMYPLQ WQLINKGCPT DETVLVHENG KDHRATFQFN AFRFQNIPKL SKVWLHCETF ICDSEKLSCP VNCDKRKRML RDQTGGVLVV ELSLRSRA
Greater than 83.7% as determined by SDS-PAGE.
21684
100 μg
Not determined.
Full Length of Mature Protein
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers