missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Serum amyloid A-2 Protein

A DNA sequence encoding the Mus musculus (Mouse) Serum amyloid A-2 protein, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6649-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16064810

  • 2985.00 EUR / 1mg
Estimated Shipment: 02-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_035444.1
Serum amyloid A-2 Protein
Research Use Only
Saa2
Recombinant Protein
E. coli
GFFSFIGEAF QGAGDMWRAY TDMKEAGWKD GDKYFHARGN YDAAQRGPGG VWAAEKISDA RESFQEFFGR GHEDTMADQE ANRHGRSGKD PNYYRPPGLP AKY
Greater than 90% as determined by SDS-PAGE.
20209
1 mg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers