missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Selenoprotein 1
A DNA sequence encoding the Mus musculus (Mouse) Selenoprotein T, was expressed in the hosts and tags indicated.
245.00 EUR - 2985.00 EUR
Specifications
Accession Number | NP_001035486.2 |
---|---|
Gene ID (Entrez) | 69227 |
Name | Selenoprotein 1 |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16057850
|
enQuireBio™
QP7250-EC-100UG |
100 μg |
675.00 EUR
100µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16077850
|
enQuireBio™
QP7250-EC-1MG |
1 mg |
2985.00 EUR
1mg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16067850
|
enQuireBio™
QP7250-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16087850
|
enQuireBio™
QP7250-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16097850
|
enQuireBio™
QP7250-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16007860
|
enQuireBio™
QP7250-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001035486.2 | |
Selenoprotein 1 | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
69227 | |
Research Use Only | |
Selt | |
Recombinant Protein | |
E. coli | |
SANLGGVPSK RLKMQYATGP LLKFQICVSU GYRRVFEEYM RVISQRYPDI RIEGENYLPQ PIYRHIASFL SVFKLVLIGL IIVGKDPFAF FGMQAPSIWQ WGQENKVYAC MMVFFLSNMI ENQCMSTGAF EITLNDVPVW SKLESGHLPS MQQLVQILDN EMKLNVHMDS IPHHRS | |
Greater than 90% as determined by SDS-PAGE. |