missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse S100A8 / CAGA Protein
A DNA sequence encoding the Mus musculus (Mouse) S100A8 / CAGA, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6641-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_038678.1 | |
S100A8 / CAGA Protein | |
Research Use Only | |
S100A8 | |
Recombinant Protein | |
E. coli | |
PSELEKALSN LIDVYHNYSN IQGNHHALYK NDFKKMVTTE CPQFVQNINI ENLFRELDIN SDNAINFEEF LAMVIKVGVA SHKDSHKE | |
Greater than 90% as determined by SDS-PAGE. |
20201 | |
10 μg | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |