missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse REG3G / PAP1B Protein

A DNA sequence encoding the Mus musculus (Mouse) REG3G / PAP1B, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6595-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16024420

  • 1925.00 EUR / 500µg
Estimated Shipment: 28-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_035390.1
REG3G / PAP1B Protein
Research Use Only
Reg3g
Recombinant Protein
Yeast
EVAKKDAPSS RSSCPKGSRA YGSYCYALFS VSKNWYDADM ACQKRPSGHL VSVLSGAEAS FLSSMIKSSG NSGQYVWIGL HDPTLGYEPN RGGWEWSNAD VMNYINWETN PSSSSGNHCG TLSRASGFLK WRENYCNLEL PYVCKFKA
Greater than 90% as determined by SDS-PAGE.
19695
500 μg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers