missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse REG3G / PAP1B Protein
A DNA sequence encoding the Mus musculus (Mouse) REG3G / PAP1B, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6595-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_035390.1 | |
REG3G / PAP1B Protein | |
Research Use Only | |
Reg3g | |
Recombinant Protein | |
E. coli | |
EVAKKDAPSS RSSCPKGSRA YGSYCYALFS VSKNWYDADM ACQKRPSGHL VSVLSGAEAS FLSSMIKSSG NSGQYVWIGL HDPTLGYEPN RGGWEWSNAD VMNYINWETN PSSSSGNHCG TLSRASGFLK WRENYCNLEL PYVCKFKA | |
Greater than 90% as determined by SDS-PAGE. |
19695 | |
50 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |