missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Ras-related protein Rab-10 Protein
A DNA sequence encoding the Mus musculus (Mouse) Ras-related protein Rab-10, was expressed in the hosts and tags indicated.
245.00 EUR - 2985.00 EUR
Specifications
Accession Number | NP_057885.1 |
---|---|
Gene ID (Entrez) | 19325 |
Name | Ras-related protein Rab-10 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16014210
|
enQuireBio™
QP6565-EC-100UG |
100 μg |
675.00 EUR
100µg |
Estimated Shipment: 26-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16034210
|
enQuireBio™
QP6565-EC-1MG |
1 mg |
2985.00 EUR
1mg |
Estimated Shipment: 26-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16024210
|
enQuireBio™
QP6565-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16044210
|
enQuireBio™
QP6565-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16054210
|
enQuireBio™
QP6565-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16064210
|
enQuireBio™
QP6565-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_057885.1 | |
Ras-related protein Rab-10 Protein | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
19325 | |
Research Use Only | |
Rab10 | |
Recombinant Protein | |
E. coli | |
MAKKTYDLLF KLLLIGDSGV GKTCVLFRFS DDAFNTTFIS TIGIDFKIKT VELQGKKIKL QIWDTAGQER FHTITTSYYR GAMGIMLVYD ITNGKSFENI SKWLRNIDEH ANEDVERMLL GNKCDMDDKR VVPKGKGEQI AREHGIRFFE TSAKANINIE KAFLTLAEDI LRKTPVKEPN SENVDISSGG GVTGWKSKCC | |
Greater than 90% as determined by SDS-PAGE. |