missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Protein FAM83D Protein

A DNA sequence encoding the Mus musculus (Mouse) Protein FAM83D, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9893-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16028941

  • 810.00 EUR / 100µg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_082251.2
Protein FAM83D Protein
Research Use Only
Fam83d
Recombinant Protein
Yeast
MAARFELLDD LPAACLSPCG PPNPTELFSE ARRLALEQLL AGGPDAWAAF LRRERLGRFL NADEVREVLG AAERPGEDGA AVAEDSFGSS HECSSGTYFP EQSDLEPPAL ELGWPSFYQG AYRGATRVEA HFQPRGAGAG GPYGCKDALR QQLRSAREVI AVVMDVFSDI DIFRDLQESC RKRGVAVYIL LDQTLLPHFL DMCMDLRVHP EQEKLMTVRT ITGNIYYARS GTKVVGKVHE KFTLIDGIRV ATGSYSFTWT DGKLNSSNLV ILSGQVVEHF DLEFRILYAQ SEPISSKLLS NFQINSKFDH LADRKPQSKE PTLGNLLRMR LARLSSTPRK SNLGPEEPPK DRAKPKRPDS EASTISDEDY FHSHKDQLED SKVADAATQT EPREEMAAVS LSEVGTQTSS SMMCVGTQTT VVTRAASSQA TVWSKSTTTQ TEADESFLPQ GAQSKEGSPA SKMSVSRSSS VRSSSSVSSQ GSLASSVSSH VSLTAADLHT PAYPKYLGLG TPHLDLCLRD SFRNLSKERQ VHFTGIRSRL TQMLTVLSRR TLFTEHYLSY SPGSFTRAST NLVSVRDIAL YPPYQ
Greater than 90% as determined by SDS-PAGE.
71878
100 μg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers