missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Protein FAM83D Protein
A DNA sequence encoding the Mus musculus (Mouse) Protein FAM83D, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9893-ye-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_082251.2 | |
Protein FAM83D Protein | |
Research Use Only | |
Fam83d | |
Recombinant Protein | |
Yeast | |
MAARFELLDD LPAACLSPCG PPNPTELFSE ARRLALEQLL AGGPDAWAAF LRRERLGRFL NADEVREVLG AAERPGEDGA AVAEDSFGSS HECSSGTYFP EQSDLEPPAL ELGWPSFYQG AYRGATRVEA HFQPRGAGAG GPYGCKDALR QQLRSAREVI AVVMDVFSDI DIFRDLQESC RKRGVAVYIL LDQTLLPHFL DMCMDLRVHP EQEKLMTVRT ITGNIYYARS GTKVVGKVHE KFTLIDGIRV ATGSYSFTWT DGKLNSSNLV ILSGQVVEHF DLEFRILYAQ SEPISSKLLS NFQINSKFDH LADRKPQSKE PTLGNLLRMR LARLSSTPRK SNLGPEEPPK DRAKPKRPDS EASTISDEDY FHSHKDQLED SKVADAATQT EPREEMAAVS LSEVGTQTSS SMMCVGTQTT VVTRAASSQA TVWSKSTTTQ TEADESFLPQ GAQSKEGSPA SKMSVSRSSS VRSSSSVSSQ GSLASSVSSH VSLTAADLHT PAYPKYLGLG TPHLDLCLRD SFRNLSKERQ VHFTGIRSRL TQMLTVLSRR TLFTEHYLSY SPGSFTRAST NLVSVRDIAL YPPYQ | |
Greater than 90% as determined by SDS-PAGE. |
71878 | |
50 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |