missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Prealbumin / Transthyretin / TTR / PALB Protein
A DNA sequence encoding the Mus musculus (Mouse) Prealbumin / Transthyretin / TTR / PALB, was expressed in the hosts and tags indicated.
287.00 EUR - 3575.00 EUR
Specifications
Accession Number | NP_038725.1 |
---|---|
Gene ID (Entrez) | 22139 |
Name | Prealbumin / Transthyretin / TTR / PALB Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16086270
|
enQuireBio™
QP6849-YE-100UG |
100 μg |
810.00 EUR
100µg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16006280
|
enQuireBio™
QP6849-YE-1MG |
1 mg |
3575.00 EUR
1mg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16096270
|
enQuireBio™
QP6849-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16016280
|
enQuireBio™
QP6849-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16026280
|
enQuireBio™
QP6849-YE-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16036280
|
enQuireBio™
QP6849-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_038725.1 | |
Prealbumin / Transthyretin / TTR / PALB Protein | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
22139 | |
Research Use Only | |
TTR | |
Recombinant Protein | |
Yeast | |
GPAGAGESKC PLMVKVLDAV RGSPAVDVAV KVFKKTSEGS WEPFASGKTA ESGELHGLTT DEKFVEGVYR VELDTKSYWK TLGISPFHEF ADVVFTANDS GHRHYTIAAL LSPYSYSTTA VVSNPQN | |
Greater than 90% as determined by SDS-PAGE. |