missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Placenta-expressed transcript 1 Protein
A DNA sequence encoding the Mus musculus (Mouse) Placenta-expressed transcript 1 protein, was expressed in the hosts and tags indicated.
245.00 EUR - 2985.00 EUR
Specifications
Accession Number | NP_083915.2 |
---|---|
Gene ID (Entrez) | 76509 |
Name | Placenta-expressed transcript 1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16011931
|
enQuireBio™
QP7974-EC-100UG |
100 μg |
675.00 EUR
100µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16031931
|
enQuireBio™
QP7974-EC-1MG |
1 mg |
2985.00 EUR
1mg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16021931
|
enQuireBio™
QP7974-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16041931
|
enQuireBio™
QP7974-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16051931
|
enQuireBio™
QP7974-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16061931
|
enQuireBio™
QP7974-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_083915.2 | |
Placenta-expressed transcript 1 Protein | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
76509 | |
Research Use Only | |
Plet1 | |
Recombinant Protein | |
E. coli | |
SDNGSCVVLD NIYTSDILEI STMANVSGGD VTYTVTVPVN DSVSAVILKA VKEDDSPVGT WSGTYEKCND SSVYYNLTSQ SQSVFQTNWT VPTSEDVTKV NLQVLIVVNR TASKSSVKME QVQPSASTPI PESSETSQTI NTTPTVNTAK TTAKDTANTT AVTTANTTAN TTAVTTAKTT AKSLAIRTLG S | |
Greater than 86.6% as determined by SDS-PAGE. |