missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Pcsk5 Protein
A DNA sequence encoding the Mus musculus (Mouse) Pcsk5, was expressed in the hosts and tags indicated.
245.00 EUR - 2985.00 EUR
Specifications
Accession Number | NP_001177412.1 |
---|---|
Gene ID (Entrez) | 18552 |
Name | Pcsk5 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16003560
|
enQuireBio™
QP6474-EC-100UG |
100 μg |
675.00 EUR
100µg |
Estimated Shipment: 26-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16023560
|
enQuireBio™
QP6474-EC-1MG |
1 mg |
2985.00 EUR
1mg |
Estimated Shipment: 26-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16013560
|
enQuireBio™
QP6474-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16033560
|
enQuireBio™
QP6474-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16043560
|
enQuireBio™
QP6474-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16053560
|
enQuireBio™
QP6474-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001177412.1 | |
Pcsk5 Protein | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
18552 | |
Research Use Only | |
Pcsk5 | |
Recombinant Protein | |
E. coli | |
DYDLSHAQST YFNDPKWPSM WYMHCSDNTH PCQSDMNIEG AWKRGYTGKN IVVTILDDGI ERTHPDLMQN YDALASCDVN GNDLDPMPRY DASNENKHGT RCAGEVAATA NNSHCTVGIA FNAKIGGVRM LDGDVTDMVE AKSVSYNPQH VHIYSASWGP DDDGKTVDGP APLTRQAFEN GVRMGRRGLG SVFVWASGNG GRSKDHCSCD GYTNSIYTIS ISSTAESGKK PWYLEECSST LATTYSSGES YDKKIITTDL RQRCTDNHTG TSASAPMAAG IIALALEANP FLTWRDVQHV IVRTSRAGHL NANDWKTNAA GFKVSHLYGF GLMDAE | |
Greater than 90% as determined by SDS-PAGE. |