missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse p53 / TP53 Protein

A DNA sequence encoding the Mus musculus (Mouse) p53 / TP53, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6823-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16006090

Quantity Price
1 245.00 EUR / 10µg
6 228.51 EUR / 10µg
Estimated Shipment
03-09-2020
Add to basket


Specifications

Specifications

NP_001120705.1
p53 / TP53 Protein
10 μg
Not determined.
Full Length
Mouse
His-SUMO
Tris-based buffer, 50% glycerol
22059
Greater than 90% as determined by SDS-PAGE.
Research Use Only
TP53
Recombinant Protein
E. coli
MEESQSDISL ELPLSQETFS GLWKLLPPED ILPSPHCMDD LLLPQDVEEF FEGPSEALRV SGAPAAQDPV TETPGPVAPA PATPWPLSSF VPSQKTYQGN YGFHLGFLQS GTAKSVMCTY SPPLNKLFCQ LAKTCPVQLW VSATPPAGSR VRAMAIYKKS QHMTEVVRRC PHHERCSDGD GLAPPQHLIR VEGNLYPEYL EDRQTFRHSV VVPYEPPEAG SEYTTIHYKY MCNSSCMGGM NRRPILTIIT LEDSSGNLLG RDSFEVRVCA CPGRDRRTEE ENFRKKEVLC PELPPGSAKR ALPTCTSASP PQKKKPLDGE YFTLKIRGRK RFEMFRELNE ALELKDAHAT EESGDSRAHS SYLKTKKGQS TSRHKKTMVK KVGPDSD
Videos
Documents