missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Muellerian-inhibiting factor Protein
A DNA sequence encoding the Mus musculus (Mouse) Muellerian-inhibiting factor, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8974-ye-200ug
Additional Details : Weight : 0.01000kg
Specifications
Muellerian-inhibiting factor Protein | |
Research Use Only | |
AMH | |
Recombinant Protein | |
Yeast | |
DKGQDGPCAL RELSVDLRAE RSVLIPETYQ ANNCQGACRW PQSDRNPRYG NHVVLLLKMQ ARGAALGRLP CCVPTAYAGK LLISLSEERI SADHVPNMVA TEC | |
Greater than 90% as determined by SDS-PAGE. |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |