missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Muellerian-inhibiting factor Protein

A DNA sequence encoding the Mus musculus (Mouse) Muellerian-inhibiting factor, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8974-ye-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16017351

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

Muellerian-inhibiting factor Protein
Research Use Only
AMH
Recombinant Protein
Yeast
DKGQDGPCAL RELSVDLRAE RSVLIPETYQ ANNCQGACRW PQSDRNPRYG NHVVLLLKMQ ARGAALGRLP CCVPTAYAGK LLISLSEERI SADHVPNMVA TEC
Greater than 90% as determined by SDS-PAGE.
10 μg
Not determined.
Partial (see sequence information for more details).
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers