missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse MUC5AC Protein

A DNA sequence encoding the Mus musculus (Mouse) MUC5AC, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8891-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16087011

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_034974.1
MUC5AC Protein
Research Use Only
Muc5ac
Recombinant Protein
E. coli
CPKNSSLIVT YEEGACCPTQ NCSSQKGCEV NGTLYQPGDV VSSSLCERCL CEVSSNPLSD VFMVSCETEL CNTQCPKGSE YQAMPGQCCG KCIPKTCPFK NNSGSTYFYQ PGELWAEPGN PCVTHKCEKF QDVLMVVTMK TECPKINCPQ GQAQLREDGC CYDCPLPNQQ KCTVHQRQQI IRQQNCSSEG PVSISYCQGN CGDSISMYSL EANKVEHTCE CCQELQTSQR NVTLRCDDGS SQTFSYTQVE KCGCLGQQCH ALGDTSHAES
Greater than 90% as determined by SDS-PAGE.
17833
10 μg
Not determined.
Partial (see sequence information for more details).
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions